missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS™ Glass Bodied Combination pH Electrode

Product Code. 10282921
Click to view available options
Connector Types:
BNC
Screw Cap
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10282921

Brand: Thermo Scientific™ 8102SC

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Capture accurate measurements of a variety of sample types, including solvents, with Thermo Scientific™ Orion™ glass body pH electrodes.

Thermo Scientific™ Orion™ ROSS Refillable Electrodes provide optimum accuracy and stability while having the additional advantages of a double junction design. They are available in chemically resistant glass bodies and best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries.
 

Features:

  • Works in extreme case of samples varying from one another by 50°C or more
  • Drift is less than 0.002 pH per day, restandardization is minimized
  • General purpose, top performance combination electrode
  • Double junction
  • TRIS compatible
  • Waterproof BNC connector

Includes:

  • One bottle 810007 fill solution

Requires:

  • pH buffers for calibration
  • Beakers, plastic or glass
  • Magnetic stirrer or Orion stirrer probe
  • Distilled or deionized water

Recommended for:

  • Quality Control (QC) and research applications

TRUSTED_SUSTAINABILITY

Specifications

Connector Types Screw Cap
Junction Ceramic
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination pH Electrode, screw cap connector
Drift <0.002 pH/day
Filling Solution Type 3M KCl
For Use With (Equipment) Standard pH Meter, with separate cable with screw cap connector
Includes One 60mL bottle of Orion ROSS filling solution and spout cap
Internal Reference ROSS
Precision 0.01
Storage Requirements Short-term Storage (up to one week): Soak the electrode in ROSS' pH electrode storage solution, Cat. No. 810001, or standard pH electrode storage solution, Cat. No. 910001. For enhanced electrode performance, use ROSS' pH electrode storage solution. Long-term Storage (more than one week): Fill the reference chamber and securely cover the fill hole. Cover the sensing bulb and reference junction with the protective cap, sleeve or storage bottle containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode.
Temperature (English) Operating 32°F to 212°F
Length (English) 4.72 in.
Length (English) Cable 39.37 in.
Length (Metric) 120 mm
Length (Metric) Cable 1 m
Diameter (English) Body 0.47 in.
Diameter (Metric) Body 12 mm
Diameter (Metric) Cap 16 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.