Learn More
Thermo Scientific™ Orion™ AQUAfast 24mm Round Vials for Colorimeters and Spectrophotometers
Use Thermo Scientific™ Orion™ AQUAfast 24mm Round Vials with a variety of Orion™ colorimeters and spectrophotometers, each package contains 12 vials
Brand: Thermo Scientific™ AC2V24
Code : A0
Additional Details : Weight : 0.01000kg
Description
Use Thermo Scientific™ Orion™ AQUAfast 24mm Round Vials with a variety of Orion™ colorimeters and spectrophotometers. The Orion™ AQUAfast 24mm Round Vials can be used with Orion™ AQUAfast colorimetric powder pack, tablet and liquid reagents to measure a variety of common water and wastewater parameters. Catalog number AC2V24 contains 12 x 24mm vials.
Specifications
Round vials | |
12 Cs. | |
Vial |
24 mm | |
Certificate of Analysis (COA) |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.