Learn More
ABEBA™ Original Plus Black Occupational Clogs OB
Product Code. 16348191
Click to view available options
Size:
36
37
38
39
40
41
42
Unit Size:
1 pair
Product Code. 16348191
Brand: ABEBA™ 37531/36
Only null left
Add to basket
Add to basket
ABEBA™ Original Plus Black Occupational Clogs OB are washable at 30 degrees and feature a foldable heel strap and slip resistant TPU outsole.
  • Quality, comfort, durability
  • Comply to EN ISO 20347 : 2012, OB SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water and chemical microfiber
  • Foldable heel strap, adjustable
  • Sanitized treatment
Specifications
Buckle Material Stainless steel
Certifications/Compliance CE EN ISO 20347 :2012, OB, A, E, FO, SRC, ESD
Closure Type Heel foldable strap
Color Black
Material Microfiber
Size 36
Sole Features Slip and abrasion resistant, ESD dissipative, shock absorbent
Sole Material Thermoplastic Polyurethane
Style Clogs
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) EN ISO 20347 : 2012; DIN EN 61340
Resistant To Water