missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Oridonin

Product Code. 16471476
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471476 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471476 Supplier MedChemExpress Supplier No. HYN0004
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Oridonin (NSC-250682), a diterpenoid isolated from Rabdosia rubescens, acts as an inhibitor of AKT, with IC50s of 8.4 and 8.9 μM for AKT1 and AKT2; Oridonin possesses anti-tumor, anti-bacterial and anti-inflammatory effects.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 364.43 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 28957-04-2
Product Type Natural Products
Purity 98.86%
Solubility DMSO : 125 mg/mL (343.00 mM∣ultrasonic and warming and heat to 60°C)
Synonym NSC-250682∣ Isodonol
Target Akt∣Bacterial
Quantity 200 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.