missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Oridonin

Product Code. 16438818
Encompass_Preferred
Click to view available options
:
10 mg
This item is not returnable. View return policy

Product Code. 16438818

Brand: MedChemExpress HYN0004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oridonin (NSC-250682), a diterpenoid isolated from Rabdosia rubescens, acts as an inhibitor of AKT, with IC50s of 8.4 and 8.9 μM for AKT1 and AKT2; Oridonin possesses anti-tumor, anti-bacterial and anti-inflammatory effects.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 364.43 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 28957-04-2
Product Type Natural Products
Purity 98.86%
Solubility DMSO : 125 mg/mL (343.00 mM∣ultrasonic and warming and heat to 60°C)
Synonym NSC-250682∣ Isodonol
Target Akt∣Bacterial
Quantity 10 mg
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.