missing translation for 'onlineSavingsMsg'
Learn More

Organophosphorus Pesticides Mixture 597 100 μg/mL in Ethyl acetate, Dr. Ehrenstorfer

Product Code. 30309843
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309843 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309843 Supplier Dr. Ehrenstorfer Supplier No. DREA50000597EA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acephate∣Diazinon∣Dichlorvos∣Dimethoate∣Ethion∣Malathion∣Methamidophos∣Methidathion∣Monocrotophos∣Omethoate∣Parathion-ethyl∣Parathion-methyl
CAS 30560-19-1, 333-41-5, 62-73-7, 60-51-5, 563-12-2, 121-75-5
Analyte or Component Names Acephate∣Diazinon∣Dichlorvos∣Dimethoate∣Ethion∣Malathion∣Methamidophos∣Methidathion∣Monocrotophos∣Omethoate∣Parathion-ethyl∣Parathion-methyl
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Ethyl Acetate
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Acephate 100 μg/mL∣Diazinon 100 μg/mL∣Dichlorvos 100 μg/mL∣Dimethoate 100 μg/mL∣Ethion 100 μg/mL∣Malathion 100 μg/mL∣Methamidophos 100 μg/mL∣Methidathion 100 μg/mL∣Monocrotophos 100 μg/mL∣Omethoate 100 μg/mL∣Parathion-ethyl 100 μg/mL∣Parathion-methyl 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.