Learn More
Description
The Organoid Essentials Package contains the key components required for the development of organoid models in cell culture.
The package contains the following five vials:
- 10 μg Mouse Wnt-3a (Cat. No. 315-20-10UG)
- 100 μg Human Noggin (Cat. No. 120-10C-100UG)
- 100 μg Human R-Spondin 1 (Cat. No. 120-38-100UG)
- 100 μg Human FGF-10 (Cat. No. 100-26-100UG)
- 100 μg Animal-Free Human EGF (Cat. No. AF-100-15-100UG)
FGF-10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. FGF-10 plays a crucial role in epithelial-mesenchymal interactions, which are essential for the development of various organs.
EGF (epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli or to EGF binding to the receptor. EGF promotes the rapid proliferation of epithelial progenitor cells, which is essential for the initial growth and expansion of organoids.
R-Spondin-1 is a member of the R-spondin family and encodes a secreted activator protein with two cysteine-rich, furin-like domains and one thrombospondin type 1 domain.
Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Wnt-3A supports the growth and morphogenesis of organoids by encouraging the proliferation and differentiation of progenitor cells into various cell types, contributing to the development of complex tissue structures.
Specifications
Specifications
| Format | Lyophilized |
| Product Type | Stem Cell Expansion Cytokine Package |
| Quantity | 1 Ea. |
| Species | Human, Murine |
For Research Use Only.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.