missing translation for 'onlineSavingsMsg'
Learn More

Organochlorine Pesticides Mixture 942 1000 μg/mL in Methyl tert-butyl ether, Dr. Ehrenstorfer

Product Code. 30328096
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328096 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328096 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000942MB
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Methoxychlor
CAS 72-54-8, 72-55-9, 50-29-3, 309-00-2, 959-98-8, 319-84-6
Analyte or Component Names 4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Methoxychlor
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix MTBE
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 4,4'-DDD 1000 μg/mL∣4,4'-DDE 1000 μg/mL∣4,4'-DDT 1000 μg/mL∣Aldrin 1000 μg/mL∣alpha-Endosulfan 1000 μg/mL∣alpha-HCH 1000 μg/mL∣beta-Endosulfan 1000 μg/mL∣beta-HCH 1000 μg/mL∣delta-HCH 1000 μg/mL∣Dieldrin 1000 μg/mL∣Endosulfan-sulfate 1000 μg/mL∣Endrin 1000 μg/mL∣Endrin-aldehyde 1000 μg/mL∣gamma-HCH 1000 μg/mL∣Heptachlor 1000 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 1000 μg/mL∣Methoxychlor 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.