missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organochlorine Pesticides Mixture 637 500 μg/mL in Hexane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣delta-HCH∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Isodrin∣Methoxychlor∣Mirex |
| CAS | 53-19-0, 3424-82-6, 789-02-6, 72-54-8, 72-55-9, 50-29-3 |
| Analyte or Component Names | 2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣delta-HCH∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Isodrin∣Methoxychlor∣Mirex |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hexane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 2,4'-DDD 500 μg/mL∣2,4'-DDE 500 μg/mL∣2,4'-DDT 500 μg/mL∣4,4'-DDD 500 μg/mL∣4,4'-DDE 500 μg/mL∣4,4'-DDT 500 μg/mL∣Aldrin 500 μg/mL∣alpha-Endosulfan 500 μg/mL∣alpha-HCH 500 μg/mL∣beta-Endosulfan 500 μg/mL∣beta-HCH 500 μg/mL∣delta-HCH 500 μg/mL∣Dieldrin 500 μg/mL∣Endrin 500 μg/mL∣gamma-HCH 500 μg/mL∣Heptachlor 500 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 500 μg/mL∣Hexachlorobenzene 500 μg/mL∣Isodrin 500 μg/mL∣Methoxychlor 500 μg/mL∣Mirex 500 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?