Learn More
Thermo Scientific™ Torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Product Code. 13552790 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
ICP-MS Quartz Torch
Unit Size:
Each
Product Code. Type unitSize
13552790 ICP-MS Quartz Torch Each
1 options
Product Code. 13552790 Supplier Thermo Scientific™ Supplier No. 1230790
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Get improved plasma stability from the tulip design and easy, push-in insertion of Thermo Scientific™ torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems.
Gain improved plasma stability from the tulip design and easy, push-in Gain improved plasma stability from the tulip design and easy, push-in insertion of fully demountable, single piece, Thermo Scientific™ Torches with connection-free gas supply for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems. All connectivity (argon gas supplies and plasma ignition) is contained within the torch holder to reduce complexity and improve usability.
Specifications
Type ICP-MS Quartz Torch
For Use With (Equipment) iCAP MX and iCAP Q/Qnova Series ICP-MS Systems