missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Organic phosphorus 3 Monoclonal Antibody (2G11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549667
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
Organic phosphorus is an essential nutrient for plants and is commonly found in organic matter, such as decaying plant and animal material. Organic phosphorus 3n is a specific gene target that encodes for a protein involved in the uptake and utilization of organic phosphorus in plants. This gene is important for plant growth and development, as well as for the overall health and productivity of agricultural crops.
Specifications
| Organic phosphorus 3 | |
| Monoclonal | |
| 2.6 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Conjugate of OP-III hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 2G11 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction