Learn More
KERN and SOHN Organ Scale
Product Code. 17848858
Encompass_Preferred
Change view
Click to view available options
Capacity:
5 to 7.5 kg
Product Code. Capacity
17848858 5 to 7.5 kg
1 options
Product Code. 17848858 Supplier KERN and SOHN Supplier No. FOB7K4NLO
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Compact stainless steel scale, well protected with IP67 protection, and hygienic to clean
  • Particularly suitable to determine the specific weight of organs, tissues, etc. in pathology or in the laboratory
  • The stainless steel design of the housing and platform, with smooth surface, makes the scale rust-free and easy to clean
  • Protection against dust and water splashes IP67 (only when using battery)
  • High mobility: thanks to battery operation, compact, lightweight construction, it is suitable for the use in several locations
  • Protective working cover included with delivery
Specifications
Type Organ Scale
Capacity 5 to 7.5 kg
Display Type LCD