missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORF73/HHV8 Rat anti-Nonspecies specific, Clone: LN53, Novus Biologicals™
Rat Monoclonal Antibody
£405.00
Specifications
| Antigen | ORF73/HHV8 |
|---|---|
| Clone | LN53 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA 1-5 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
ORF73/HHV8 Monoclonal specifically detects ORF73/HHV8 in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ORF73/HHV8 | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
| 10 mM PBS with 0.05% BSA | |
| 935114 | |
| IgG2c κ | |
| Protein A or G purified |
| LN53 | |
| ELISA 1-5 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV | |
| Recombinant protein corresponding to the latent nuclear antigen 1 molecule of ORF73/HHV8 | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title