missing translation for 'onlineSavingsMsg'
Learn More

ORF73/HHV8 Mouse anti-Nonspecies specific, Clone: HHV8/3606, Novus Biologicals™

Product Code. 18339991 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339991 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339991 Supplier Novus Biologicals Supplier No. NBP30780920UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ORF73/HHV8 Monoclonal specifically detects ORF73/HHV8 in Non-species specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORF73/HHV8
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HHV8/3606
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA 1-5 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Formulation 10 mM PBS with 0.05% BSA
Gene Alias HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV
Host Species Mouse
Immunogen Recombinant protein corresponding to the latent nuclear antigen 1 molecule of ORF73/HHV8
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 935114
Target Species Non-species specific
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.