missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORF73/HHV8 Antibody (LN53), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-08217H
This item is not returnable.
View return policy
Description
ORF73/HHV8 Monoclonal antibody specifically detects ORF73/HHV8 in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ORF73/HHV8 | |
| Monoclonal | |
| HRP | |
| HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV | |
| Recombinant protein corresponding to the latent nuclear antigen 1 molecule of ORF73/HHV8 | |
| 100 μg | |
| Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
| 935114 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| LN53 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction