missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORF73/HHV8 Antibody (LN53), DyLight 350, Novus Biologicals™
Description
ORF73/HHV8 Monoclonal antibody specifically detects ORF73/HHV8 in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Specifications
Specifications
| Antigen | ORF73/HHV8 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay |
| Classification | Monoclonal |
| Clone | LN53 |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV |
| Host Species | Rat |
| Immunogen | Recombinant protein corresponding to the latent nuclear antigen-1 molecule of HHV8. |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?