missing translation for 'onlineSavingsMsg'
Learn More

ORF73/HHV8 Antibody (LN53), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18430243 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430243 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430243 Supplier Novus Biologicals Supplier No. NBP122767AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

ORF73/HHV8 Monoclonal antibody specifically detects ORF73/HHV8 in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORF73/HHV8
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Classification Monoclonal
Clone LN53
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias HHV 8, Human Herpes Virus 8, Kaposi's sarcoma associated herpes virus, KSHV
Host Species Rat
Immunogen Recombinant protein corresponding to the latent nuclear antigen-1 molecule of HHV8.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 935114
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.