missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Orexin Receptor 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584204
This item is not returnable.
View return policy
Description
Immunogen sequence: WCRQIPGTSS VVQRKWKPLQ PVSQPRGPGQ PTKSRMSAVA AEIKQIRARR K.
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein binds the hypothalamic neuropeptides orexin A and orexin B. A related gene encodes a G-protein coupled receptor that selectively binds orexin A.
Specifications
| Orexin Receptor 2 | |
| Polyclonal | |
| Unconjugated | |
| HCRTR2 | |
| HCRTR1; Hcrtr2; hypocretin (orexin) receptor 2; hypocretin receptor 2; hypocretin receptor type 2; mOX2aR; mOX2bR; Mox2r; mOXR2; orexin receptor 1; orexin receptor 2; orexin receptor type 2; orexin receptor-1; orexin receptor-2; orexin type-2 receptor; OTTHUMP00000039969; OX1R; ox2r; ox-2-R; ox2-R | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3062 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43614 | |
| HCRTR2 | |
| Recombinant protein corresponding to Human HCRTR2. Recombinant protein control fragment (Product #RP-101312). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction