missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Orexin Receptor 2 Monoclonal Antibody (456738)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524178
This item is not returnable.
View return policy
Description
This antibody stains human Orexin R2/HCRTR2 transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein binds the hypothalamic neuropeptides orexin A and orexin B. A related gene encodes a G-protein coupled receptor that selectively binds orexin A.
Specifications
| Orexin Receptor 2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O43614 | |
| HCRTR2 | |
| NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2 Met1-Trp444. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 456738 | |
| Unconjugated | |
| HCRTR2 | |
| HCRTR1; Hcrtr2; hypocretin (orexin) receptor 2; hypocretin receptor 2; hypocretin receptor type 2; mOX2aR; mOX2bR; Mox2r; mOXR2; orexin receptor 1; orexin receptor 2; orexin receptor type 2; orexin receptor-1; orexin receptor-2; orexin type-2 receptor; OTTHUMP00000039969; OX1R; ox2r; ox-2-R; ox2-R | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3062 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction