missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Orexin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121242
This item is not returnable.
View return policy
Description
Positive test controls include: U-251MG, Rat thymus. The target is usually found in the following locations: Cell junction, Cytoplasmic vesicle, Rough endoplasmic reticulum, synapse. Immunogen sequence: QPLPDCCRQK TCSCRLYELL HGAGNHAAGI LTL.
Neuropeptides that play a significant role in the regulation of food intake and sleep-wakefulness, possibly by coordinating the complex behavioral and physiologic responses of these complementary homeostatic functions. A broader role in the homeostatic regulation of energy metabolism, autonomic function, hormonal balance and the regulation of body fluids, is also suggested. Orexin-A binds to both OX1R and OX2R with a high affinity, whereas orexin-B binds only to OX2R with a similar high affinity.
Specifications
| Orexin | |
| Polyclonal | |
| Unconjugated | |
| HCRT | |
| HCRT; Hcrt1; Hcrt2; Hypocretin; hypocretin (orexin) neuropeptide; hypocretin neuropeptide precursor; Hypocretin-1; Hypocretin-2; NRCLP1; Orexin; orexin A; orexin B; Orexin-A; Orexin-B; OX; PPORX; Ppox; prepro-orexin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15171, 25723, 3060 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43612, O55232, O55241 | |
| HCRT | |
| A synthetic peptide corresponding to a sequence within amino acids 34-66 of human HCRT (NP_0015151). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction