missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Orexin B Recombinant Rabbit Monoclonal Antibody (2O8I1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558091
This item is not returnable.
View return policy
Description
Immunogen sequence: MNLPSTKVSW AAVTLLLLLL LLPPALLSSG AAAQPLPDCC RQKTCSCRLY ELLHGAGNHA AGILTLGKRR SGPPGLQGRL QRLLQASGNH AAGILTMGRR.
Orexin B is a neuropeptides that play a significant role in the regulation of food intake and sleep-wakefulness, possibly by coordinating the complex behavioral and physiologic responses of these complementary homeostatic functions. A broader role in the homeostatic regulation of energy metabolism, autonomic function, hormonal balance and the regulation of body fluids, is also suggested. Orexin-A binds to both OX1R and OX2R with a high affinity, whereas orexin-B binds only to OX2R with a similar high affinity.
Specifications
| Orexin B | |
| Recombinant Monoclonal | |
| 1.02 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O43612 | |
| Hcrt | |
| Synthetic peptide. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 2O8I1 | |
| Unconjugated | |
| Hcrt | |
| HCRT; Hcrt1; Hcrt2; Hypocretin; hypocretin (orexin) neuropeptide; hypocretin neuropeptide precursor; Hypocretin-1; Hypocretin-2; NRCLP1; orexin; orexin A; orexin B; orexin-A; Orexin-B; ORX; Ox; PPORX; Ppox; prepro-orexin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3060 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction