missing translation for 'onlineSavingsMsg'
Learn More

Orexin B/Hypocretin-2 Antibody (MM0501-5G21), DyLight 594, Novus Biologicals™

Product Code. 18420918 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420918 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420918 Supplier Novus Biologicals Supplier No. NBP211834DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Orexin B/Hypocretin-2 Monoclonal antibody specifically detects Orexin B/Hypocretin-2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Orexin B/Hypocretin-2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MM0501-5G21
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias HCRT, Hcrt 2, Hcrt2, Hypocretin, Hypocretin (orexin) neuropeptide precursor, Hypocretin 2, Hypocretin B, Hypocretin precursor, Hypocretin2, Orexin, Orexin neuropeptide precursor, Orexin precursor, OrexinB, PPORX, PPOX
Host Species Mouse
Immunogen Human Orexin B/Hypocretin-2 peptide
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3060
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.