missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Orexin A, Human, Bovine, Rat, Mouse

Product Code. 15426869
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426869 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426869 Supplier Thermo Scientific Chemicals Supplier No. J65378.!D
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3561.09 g/mol

appetite regulation, wakefulness, locomotor activity, hypothalamic-pituitary-adrenal activity, and pain thresholds. Orexin signaling is activated by nutrient depletion, causing an increase in food intake by delaying the signals of satiety. It activates the orexin-1 and -2 receptors with equal affinity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Species Bovine, Mouse, Rat
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 3561.09
Product Type Orexin A, human, bovine, rat, mouse
Quantity 0.5 mg
Regulatory Status RUO - research use only

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.