missing translation for 'onlineSavingsMsg'
Learn More

Orexin 2 Receptor Agonist, MedChemExpress

Product Code. 16417728
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16417728

Brand: Medchem Express HY19320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Orexin 2 Receptor Agonist is a potent (EC50 on OX2R is 23 nM) and OX2R-selective (OX1R/OX2R EC50 ratio is 70) agonist.

Orexin 2 Receptor Agonist is a potent (EC50 on OX2R is 23 nM) and OX2R-selective (OX1R/OX2R EC50 ratio is 70) agonist.

Specifications

Chemical Name or Material Orexin 2 Receptor Agonist
CAS 1796565-52-0
Color White
Purity Grade Notes Research
Molecular Formula C32H34N4O5S
Quantity 10 mg
Solubility Information DMSO : ≥ 32 mg/mL (54.54 mM)
SMILES O=C(C1=CC=CC(C)=C1)NCCNC2=CC(NS(C3=C(OC)C=CC(C4=CC(C(N(C)C)=O)=CC=C4)=C3)(=O)=O)=CC=C2
Molecular Weight (g/mol) 586.7
Formula Weight 586.7
Percent Purity 99.75%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.