missing translation for 'onlineSavingsMsg'
Learn More

Orcinol, 99.6%, MP Biomedicals™

Product Code. 11458130 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458130 50 g 50g
11438130 5 g 5g
11448130 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458130 Supplier MP Biomedicals Supplier No. 0210250650
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 63939-65-1
Molecular Formula C11H19IN2O
Molecular Weight (g/mol) 322.19
InChI Key VVLMSCJCXMBGDI-UHFFFAOYSA-M
Synonym oxotremorine m, oxotremorine-m, 3h oxotremorine-m, n,n,n-trimethyl-4-2-oxopyrrolidin-1-yl but-2-yn-1-aminium, trimethyl-4-2-oxopyrrolidin-1-yl but-2-ynyl azanium, n,n,n-trimethyl-4-2-oxo-1-pyrrolidinyl-2-butyn-1-aminium, 2-butyn-1-aminium, n,n,n-trimethyl-4-2-oxo-1-pyrrolidinyl, chembl23957, spectrum_001982, tocris-1067
PubChem CID 3027782
IUPAC Name trimethyl-[4-(2-oxopyrrolidin-1-yl)but-2-ynyl]azanium;iodide
SMILES C[N+](C)(C)CC#CCN1CCCC1=O.[I-]

Specifications

Color Tan to White
Assay Percent Range ∽ 98%
Quantity 50 g
Formula Weight 142.2
Percent Purity 99.6%
Physical Form Powder
Chemical Name or Material Orcinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.