missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC3L, Mouse anti-Human, Clone: 1G1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00023595-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: ALNNPYYYLKNEALKSEEGCIPNIAPDICIAYKLHLECSRLINLVDWSEAFATVVTAAEKMDANSATSEEMNEIIHARFIRAVSELELLGFIKPTKQKTDHVARLTWGGCSpecifications
| ORC3L | |
| Monoclonal | |
| Unconjugated | |
| origin recognition complex, subunit 3-like (yeast) | |
| LAT/LATHEO/ORC3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1G1 | |
| In 1x PBS, pH 7.4 | |
| BC035494 | |
| ORC3L | |
| ORC3L (AAH35494, 602 a.a. ∼ 711 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 23595 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction