missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC3L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | ORC3L |
|---|---|
| Dilution | Western Blot 1:200 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228889
|
Novus Biologicals
NBP3-35547-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233008
|
Novus Biologicals
NBP3-35547-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ORC3L Polyclonal antibody specifically detects ORC3L in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| ORC3L | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| IMAGE50150, LAT, LATHEOorigin recognition complex, subunit 3-like (yeast), ORC3Lhomolog of latheo, Drosophila, origin recognition complex subunit 3, Origin recognition complex subunit Latheo, origin recognition complex, subunit 3, origin recognition complex, subunit 3 (yeast homolog)-like, origin recognition complex, subunit 3 honolog, origin recognition complex, subunit 3 honolog (yeast), origin recognition complex, subunit 3-like | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human ORC3L (NP_036513.2).,, Sequence:, MATSSMSKGCFVFKPNSKKRKISLPIEDYFNKGKNEPEDSKLRFETYQLIWQQMKSENERLQEELNKNLFDNLIEFLQKSHSGFQKNSRDLGGQIKLREIPTAALVLGVNVTDHDLTFGSLTEALQNNVTPYVVSLQAKDCPDMKHFLQKLISQLMDCCVDIKSKEEESVHVTQRKTHYSMDSLSSWYMTVTQKTDPKMLSKKRTTSSQWQSPPVVVILKDMESFATKVLQDFIIISSQH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 23595 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title