missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ORC3 Polyclonal Antibody

Product Code. 11305923
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305923 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11305923 Supplier Invitrogen™ Supplier No. PA517949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Histone deacetylase 2 (HDAC2), or transcriptional regulator homolog RPD3 L1, is highly homologous to the yeast transcription factor RPD3 (reduced potassium dependency 3) gene. As in yeast, human HDA2 is likely to be involved in regulating chromatin structure during transcription. It has been implicated to associate with YY1, a mammalian zinc-finger transcription factor, which negatively regulates transcription by tethering RPD3 to DNA as a cofactor. This process is highly concerved from yeast to human.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ORC3
Gene Accession No. Q9UBD5
Gene Alias homolog of latheo, Drosophila; IMAGE50150; LAT; LATHEO; Orc3; ORC3L; origin of replication 3 homolog; origin recognition complex subunit 3; Origin recognition complex subunit Latheo; origin recognition complex, subunit 3; origin recognition complex, subunit 3 honolog; origin recognition complex, subunit 3-like; OTTHUMP00000040611; RP3-486L4.1
Gene Symbols ORC3
Host Species Goat
Immunogen Synthetic peptide sequence (QKTDHVARLTWGGC) corresponding to the C-terminus amino acids of ORC3L (aa 698-711).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23595
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.