missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC1 Mouse anti-Human, Clone: 7F6/1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07472-100ug
This item is not returnable.
View return policy
Description
ORC1 Monoclonal specifically detects ORC1 in Human samples. It is validated for ELISA.
Specifications
| ORC1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA | |
| HS, HSORC1, ORC1Lorigin recognition complex subunit 1, origin recognition complex, subunit 1, origin recognition complex, subunit 1 (yeast homolog)-like, origin recognition complex, subunit 1 homolog, origin recognition complex, subunit 1 homolog (S. cerevisiae), origin recognition complex, subunit 1-like, origin recognition complex, subunit 1-like (yeast), PARC1origin recognition complex 1, Replication control protein 1 | |
| Recombinant full-length human ORC1 protein | |
| 100 μg | |
| Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4998 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA | |
| 7F6/1 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction