missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ORC1 Monoclonal specifically detects ORC1 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | ORC1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 7F6/1 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | ELISA |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | HS, HSORC1, ORC1Lorigin recognition complex subunit 1, origin recognition complex, subunit 1, origin recognition complex, subunit 1 (yeast homolog)-like, origin recognition complex, subunit 1 homolog, origin recognition complex, subunit 1 homolog (S. cerevisiae), origin recognition complex, subunit 1-like, origin recognition complex, subunit 1-like (yeast), PARC1origin recognition complex 1, Replication control protein 1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?