missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORAOV1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549859
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ORAOV1 protein.
ORAOV1 is a protein coding gene. The complex LTO1:YAE1 functions as a target specific adapter that probably recruits apo-ABCE1 to the cytosolic iron-sulfur protein assembly (CIA) complex machinery. May be required for biogenesis of the large ribosomal subunit and initiation of translation. May play a role in the regulation of proline metabolism and ROS production.
Specifications
| ORAOV1 | |
| Polyclonal | |
| Unconjugated | |
| ORAOV1 | |
| 2210010N10Rik; Lto1; oral cancer overexpressed 1; oral cancer overexpressed protein 1-A; oral cancer-overexpressed protein 1; Oral cancer-overexpressed protein 1 homolog; Oraov1; Protein LTO1 homolog; TAOS1; Tumor-amplified and overexpressed sequence 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 220064 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WV07 | |
| ORAOV1 | |
| Synthesized peptide derived from internal of human ORAV1 (aa 81-130). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction