missing translation for 'onlineSavingsMsg'
Learn More

Orange/Orange Flexible PVC Peri-Pump Tubing, 2-Bridge, 0.89 mm (0.035 in.), 12/pack

Product Code. 30313823
Click to view available options
Color:
Orange/Orange
Material:
PVC
This item is not returnable. View return policy

Product Code. 30313823

Brand: VHG Labs VHGD180202

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 0.89 mm
Material PVC
For Use With Peristaltic Pumps
Color Orange/Orange
Flexibility Flexible
Quantity 12/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.