missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orange G, Technical Grade, TRC
Specifications
Specifica
| Chemical Name or Material | Acid Orange 10 |
| CAS | 1936-15-8 |
| Molecular Formula | C16 H10 N2 O7 S2 . 2 Na |
| Quantity | 250 mg |
| Synonym | 1,3-Naphthalenedisulfonic acid','7-hydroxy-8-(phenylazo)-','disodium salt (9CI), 2-Naphthol-6,8-disulfonic acid','1-phenylazo-','disodium salt (6CI), C.I. Acid Orange 10 (7CI), C.I. Acid Orange 10','disodium salt (8CI), 1-(Phenylazo)-2-naphthol-6,8-disulfonic acid disodium salt, 1370 Orange, Acid Crystal Orange, Acid Fast Orange EGG, Acid Fast Orange G, Acid Fast Orange GG, Acid Leather Orange G, Acid Leather Orange KG, Acid Leather Orange PGW, Acid Lightfast Orange, Acid Orange 10, Acid Orange 2G, Acid Orange 2GL, Acid Orange EGG, Acid Orange G, Acid Orange G 100, Acid Orange G Crystal, Acid Orange GG, Acid Orange Lightfast, Acidal Fast Orange, Acilan Orange GX, Amacid Crystal Orange, Amacid Light Orange G, Anadurm Orange A-G, Apocid Orange 2G, Atul Acid Crystal Orange G, Brasilan Orange 2G, Bucacid Fast Orange G, C.I. 16230, C.I. Food Orange 4, Calcocid Fast Light Orange 2G, Certicol Orange GS, Cetil Light Orange GG, Colacid Orange G, Colocid Orange 2G, Colorosacid Orange 10, Concorde Acid Orange L-GX, Crystal Orange 2G, Crystal Orange G, D and C Orange No. 3, Dinacid Crystal Orange G, Dolkwal Orange G, Duasyn Acid Orange GG, Dyacid Orange G, Dynacid Orange 2G, Egacid Orange GG, Eniacid Light Orange G, Erio Fast Orange AS, Fabracid Orange FL, Fast Acid Orange G, Fast Light Orange G, Fast Light Orange GA-CF, Fenazo Light Orange 2G, Food Orange 4, Food Orange GG, Hexacol Orange G, Hexacol Orange GG Crystals, Hidacid Fast Orange G, Hispacid Fast Orange 2G, Indacid Crystal Orange, Kemacid Crystal Orange G, Keyacid Orange 2G, Kiton Fast Orange G, Light Orange G, Multacid Orange 2G, Naphthalene Fast Orange 2G, Naphthalene Fast Orange 2GS, Naphthazine Orange 2G, Neelicol Orange G, Neklacid Fast Orange GG, Orange 2G, Orange BPC, Orange G, Orange G II, Orange GBPC, Orange GG, Pacid Orange G 10, Sandolan Fast Orange E-GL, Sandolan Orange E-GL, Solar Light Orange GX, Straight Orange G, Tertracid Light Orange G, Triacid Light Orange 2G, Vicoacid Orange IJ, Victacid Orange IJ, Vondacid Light Orange 2G, Water Orange 145924, Wool Orange 2G, Wool Orange G, Xylene Fast Orange G |
| InChI Formula | InChI=1S/C16H12N2O7S2.2Na/c19-13-7-6-10-8-12(26(20,21)22)9-14(27(23,24)25)15(10)16(13)18-17-11-4-2-1-3-5-11;;/h1-9,19H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2 |
| SMILES | [Na+].[Na+].Oc1ccc2cc(cc(c2c1N=Nc3ccccc3)S(=O)(=O)[O-])S(=O)(=O)[O-] |
| IUPAC Name | disodium;7-hydroxy-8-[(E)-phenyldiazenyl]naphthalene-1,3-disulfonate |
| Molecular Weight (g/mol) | 452.37 |
| Formula Weight | 451.9725 |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?