missing translation for 'onlineSavingsMsg'
Learn More

Melford Orange-G Loading Dye 6X Solution

Product Code. 30018642
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 30018642

Brand: Melford O2120025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

6X Concentrated loading dye for molecular biology applications.

6X Concentrated loading dye for molecular biology applications.

Specifications

Chemical Name or Material Orange-G Loading Dye 6 x Solution
CAS 7732-18-5, 1936-15-8, 56-81-5
Purity Grade Notes Molecular Biology
Quantity 25 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.