Learn More
Thermo Scientific™ Oral-Eze™ Collection Devices
Product Code. 17825878 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Oral-Eze Collection Device
Quantity:
500
Product Code. Description Quantity
17825878 Oral-Eze Collection Device 500
1 options
Product Code. 17825878 Supplier Thermo Scientific™ Supplier No. 96100500
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Take the guesswork out of collecting oral fluid samples with innovative indicator window technology.
Simplify the collection of oral samples for routine drug testing with our innovative oral fluid collection system. The Oral-Eze™ Collection Device available from Thermo Scientific™ provides all the advantages of previous collection systems with the added benefit of our indicator window technology - taking the guesswork out of sample collection.
Using the Oral-Eze Oral Fluid Collection System simplifies the collection of oral samples for routine drug testing.
  • Collection system and drug tests used with it are FDA-cleared
  • Unique indicator window turns blue when complete, taking the guesswork out of the process and reducing repeat collections
  • Fast collection time is typically 3 to 5 minutes
  • Specimen and any drugs in the specimen are stable for 21 days after collection
  • Non-negative specimens tested by the laboratory are stable for at least one year when stored frozen
TRUSTED_SUSTAINABILITY
Specifications
Description Oral-Eze Collection Device
Detectable Analytes None
Sample Type Oral Fluid
Type Collection Device
Quantity 500