missing translation for 'onlineSavingsMsg'
Learn More

ORAI3 Antibody - BSA Free, Novus Biologicals™

Product Code. 18333781 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333781 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333781 Supplier Novus Biologicals Supplier No. NBP282014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ORAI3 Polyclonal specifically detects ORAI3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORAI3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 5-10 μg/mL
Formulation PBS
Gene Alias ORAI calcium release-activated calcium modulator 3, TMEM142C
Gene Symbols ORAI3
Host Species Rabbit
Immunogen ORAI3 antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human ORAI3. The immunogen is located within amino acids 200 - 250 of ORAI3. Amino Acid Squence: ASNLPRSSASAAPSQAEPA
Molecular Weight of Antigen 34 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93129
Test Specificity ORAI3 antibody is predicted to have no cross-reactivity to ORAI1 or ORAI2.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.