missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORAI3 Antibody (2H2G9), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41326AF647
This item is not returnable.
View return policy
Description
ORAI3 Monoclonal antibody specifically detects ORAI3 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ORAI3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| ORAI calcium release-activated calcium modulator 3, TMEM142C | |
| A 19 amino acid synthetic peptide from near the carboxy terminus of human ORAI3. Amino Acid Squence: ASNLPRSSASAAPSQAEPA | |
| 0.1 mL | |
| Immunology | |
| 93129 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2H2G9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction