missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORAI3 Antibody (1B4F1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-41327PCP
This item is not returnable.
View return policy
Description
ORAI3 Monoclonal antibody specifically detects ORAI3 in Human, Rat samples. It is validated for Western Blot, ELISA
Specifications
| ORAI3 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA | |
| 1B4F1 | |
| Western Blot, ELISA | |
| ORAI calcium release-activated calcium modulator 3, TMEM142C | |
| A 19 amino acid synthetic peptide from near the carboxy terminus of human ORAI3. Amino Acid Squence: ASNLPRSSASAAPSQAEPA | |
| 0.1 mL | |
| Immunology | |
| 93129 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction