missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORAI2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565926
This item is not returnable.
View return policy
Description
Immunogen sequence: MSAELNVPIDP SAPACPEPGH KGMDYRDWVR RSYLELVTSN HHSVQALSW Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
Antigen stimulation of immune cells triggers Ca++ entry through Ca++ release-activated Ca++ (CRAC) channels. ORAI2 is one of two mammalian homologs to ORAI1, a recently identified four-transmembrane spanning protein that is an essential component of CRAC. Like ORAI1, ORAI2 has been shown to function as a highly selective Ca++ plasma membrane channel that is gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor, although at a lesser efficacy than ORAI1.
Specifications
| ORAI2 | |
| Polyclonal | |
| Unconjugated | |
| ORAI2 | |
| A730041O15Rik; C7orf19; calcium release-activated calcium channel 2; calcium release-activated calcium modulator 2 protein; CAP-binding protein complex interacting protein 2; CAP-binding protein complex-interacting protein 2; CBCIP2; H_NH0514P08.8; MEM142B; ORAI calcium release-activated calcium modulator 2; ORAI2; PP1729; Protein orai-2; putative protein ORAI2-2; RGD1310213; TMEM142B; Transmembrane protein 142B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80228 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96SN7 | |
| ORAI2 | |
| Recombinant Human ORAI2. Recombinant protein control fragment (Product #RP-106605). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction