missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orai2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-39064-25ul
This item is not returnable.
View return policy
Description
Orai2 Polyclonal specifically detects Orai2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Orai2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q96SN7 | |
| ORAI2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: IKFLPVDARRQPGPPPGPGSHTGW | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CAP-binding protein complex interacting protein 2, CAP-binding protein complex-interacting protein 2, CBCIP2C7orf19FLJ12474, chromosome 7 open reading frame 19, FLJ14733, H_NH0514P08.8, MEM142B, ORAI calcium release-activated calcium modulator 2, protein orai-2, putative protein ORAI2-2, TMEM142B, Transmembrane protein 142BFLJ44818 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 80228 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction