missing translation for 'onlineSavingsMsg'
Learn More

Orai1 Mouse anti-Human, Mouse, Rat, Janelia Fluor 646, Clone: 3F6H5, Novus Biologicals™

Product Code. 18418252 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418252 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418252 Supplier Novus Biologicals Supplier No. NBP175522JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Orai1 Monoclonal antibody specifically detects Orai1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Orai1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 3F6H5
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CRACM1ORAT1, FLJ14466, ORAI calcium release-activated calcium modulator 1, Protein orai-1, TMEM142Acalcium release-activated calcium modulator 1, Transmembrane protein 142Acalcium release-activated calcium channel protein 1
Host Species Mouse
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ORAI1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84876
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.