missing translation for 'onlineSavingsMsg'
Learn More

Orai1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen Orai1
Dilution Western Blot 1:500 - 1:1000, ELISA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30232913
View Documents
Novus Biologicals
NBP3-38425-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30228250
View Documents
Novus Biologicals
NBP3-38425-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Orai1 Polyclonal antibody specifically detects Orai1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications

Specifications

Orai1
ELISA, Western Blot
Unconjugated
Rabbit
Cancer, Neuroscience
PBS (pH 7.3), 50% glycerol
84876
IgG
Affinity purified
Western Blot 1:500 - 1:1000, ELISA
Polyclonal
Purified
RUO
Human, Mouse, Rat
CRACM1ORAT1, FLJ14466, ORAI calcium release-activated calcium modulator 1, Protein orai-1, TMEM142Acalcium release-activated calcium modulator 1, Transmembrane protein 142Acalcium release-activated calcium channel protein 1
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Orai1 (NP_116179.2).,, Sequence:, MHPEPAPPPSRSSPELPPSGGSTTSGSRRSRRRSGDGEPPGAPPPPPSAVTYPDWIGQSYSEVMSLNEHSMQALSWRKLYLSRAKLKASSRTSALLSGFA
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.