missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR8H3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571210
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 92%; Rat: 92%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
| OR8H3 | |
| Polyclonal | |
| Unconjugated | |
| OR8H3 | |
| Olfactory receptor 8H3; olfactory receptor family 8 subfamily H member 3; olfactory receptor OR11-172; olfactory receptor, family 8, subfamily H, member 3; OR11-172; OR8H3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 390152 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N146 | |
| OR8H3 | |
| synthetic peptide directed towards the C-terminal region of human OR8H3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction