missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR8D1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83343-0.1ML
This item is not returnable.
View return policy
Description
OR8D1 Polyclonal specifically detects OR8D1 in Human samples. It is validated for Western Blot.
Specifications
| OR8D1 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 283159 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| JCG9, olfactory receptor 8D1, Olfactory receptor 8D3, Olfactory receptor OR11-301, olfactory receptor, family 8, subfamily D, member 1, olfactory receptor, family 8, subfamily D, member 3, Olfactory receptor-like protein JCG9, olfactory-like receptor JCG9, OR8D3, OST004PDJ9J14 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR8D1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction