missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR7A17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571202
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 79%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 79%; Pig: 86%; Rabbit: 79%; Rat: 86%.
OR7A17 is a protein coding gene.
Specifications
| OR7A17 | |
| Polyclonal | |
| Unconjugated | |
| OR7A17 | |
| BC85395_4; HTPCRX19; Olfactory receptor 7A17; olfactory receptor family 7 subfamily A member 17; olfactory receptor family 7 subfamily A member 53; olfactory receptor OR19-20; olfactory receptor, family 7, subfamily A, member 17; olfactory receptor-like protein OLF4; OR7A17; OR7A53 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26333 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O14581 | |
| OR7A17 | |
| synthetic peptide directed towards the C-terminal region of human OR7A17. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction