missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR6X1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26089
This item is not returnable.
View return policy
Description
OR6X1 Polyclonal antibody specifically detects OR6X1 in Human samples. It is validated for Western Blot
Specifications
| OR6X1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| olfactory receptor 6X1, olfactory receptor, family 6, subfamily X, member 1, OR11-270 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human OR6X1 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 390260 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction