missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OR6M1 Polyclonal specifically detects OR6M1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | OR6M1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | olfactory receptor 6M1, Olfactory receptor OR11-271, olfactory receptor, family 6, subfamily M, member 1, olfactory receptor, family 6, subfamily M, member 2 pseudogene, OR11-271 |
| Gene Symbols | OR6M1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: LRNEKVQEVLRETVNRIMTLIQRK |
| Show More |
For Research Use Only
Product Title
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?