missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR6K2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06141F
This item is not returnable.
View return policy
Description
OR6K2 Polyclonal antibody specifically detects OR6K2 in Human samples. It is validated for Western Blot
Specifications
| OR6K2 | |
| Polyclonal | |
| Western Blot | |
| olfactory receptor 6K2, Olfactory receptor OR1-17, olfactory receptor, family 6, subfamily K, member 2, OR1-17 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human OR6K2. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 81448 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction