missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR6C75 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | OR6C75 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
OR6C75 Polyclonal specifically detects OR6C75 in Human samples. It is validated for Western Blot.Specifications
| OR6C75 | |
| Polyclonal | |
| Rabbit | |
| A6NL08 | |
| 390323 | |
| Synthetic peptides corresponding to OR6C75(olfactory receptor, family 6, subfamily C, member 75) The peptide sequence was selected from the middle region of OR6C75. Peptide sequence SCIFMYIKTSARERVTLSKGVAVLNTSVAPLLNPFIYTLRNKQVKQAFKS. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| olfactory receptor 6C75, olfactory receptor, family 6, subfamily C, member 75 | |
| OR6C75 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title