missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR6C1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | OR6C1 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
OR6C1 Polyclonal specifically detects OR6C1 in Human samples. It is validated for Western Blot.Specifications
| OR6C1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| olfactory receptor, family 6, subfamily C, member 1, OST267olfactory receptor 6C1 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR6C1 (NP_001005182). Peptide sequence PSTSQRTKAFSTCSSHMVVVSISYGSCIFMYIKPSAKDRVSLSKGVAILN | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 390321 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel